Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HHAT
Entrez Gene ID
55733
Gene Name
hedgehog acyltransferase
Gene Aliases
MART2, SKI1, Skn
Chromosome Location
Chr.1: 210327873 - 210676298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122834.3 NP_001116306.1 6 - 7 862 59
NM_001170580.2 NP_001164051.1 6 - 7 917 59
NM_001170587.2 NP_001164058.1 5 - 6 794 59
NM_001170588.2 NP_001164059.1 5 - 6 733 59
NM_018194.5 NP_060664.2 6 - 7 928 59
XM_006711441.3 XP_006711504.1 7 - 8 989 59
XM_011509737.2 XP_011508039.1 7 - 8 989 59
XM_011509738.2 XP_011508040.1 5 - 6 747 59
XM_011509739.2 XP_011508041.1 6 - 7 914 59
XM_011509740.2 XP_011508042.1 7 - 8 1118 59
XM_011509741.2 XP_011508043.1 6 - 7 862 59
XM_011509742.2 XP_011508044.1 6 - 7 771 59
XM_011509744.2 XP_011508046.1 5 - 6 922 59
XM_011509746.2 XP_011508048.1 5 - 6 748 59
XM_011509747.1 XP_011508049.1 4 - 5 712 59
XM_017001726.1 XP_016857215.1 5 - 6 963 59
XM_017001727.1 XP_016857216.1 5 - 6 963 59
XM_017001728.1 XP_016857217.1 6 - 7 1072 59
XM_017001729.1 XP_016857218.1 6 - 7 793 59
XM_017001730.1 XP_016857219.1 5 - 6 875 59
XM_017001731.1 XP_016857220.1 5 - 6 681 59
XM_017001732.1 XP_016857221.1 5 - 6 558 59
XM_017001733.1 XP_016857222.1 4 - 5 835 59
XM_017001734.1 XP_016857223.1 5 - 6 869 59
XM_017001735.1 XP_016857224.1 5 - 6 1198 59
XM_017001736.1 XP_016857225.1 5 - 6 769 59
GenBank mRNA AK001586.1 - 6 - 7 882 59
AK295130.1 - 5 - 6 814 59
AK298991.1 - 5 - 6 687 59
AK302955.1 - 5 - 6 747 59
AK316190.1 - 5 - 6 748 59
AK316524.1 - 5 - 6 780 59
AL049848.1 - 3 - 4 400 59
BC039071.1 - 6 - 7 867 59
BC117130.1 - 5 - 6 717 59

View Full Product Details