Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HHAT
Entrez Gene ID
55733
Gene Name
hedgehog acyltransferase
Gene Aliases
MART2, SKI1, Skn
Chromosome Location
Chr.1: 210327873 - 210676298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122834.3 NP_001116306.1 8 - 9 1183 93
NM_001170564.2 NP_001164035.1 6 - 7 772 93
NM_001170580.2 NP_001164051.1 8 - 9 1238 93
NM_001170587.2 NP_001164058.1 7 - 8 1115 93
NM_001170588.2 NP_001164059.1 7 - 8 1054 93
NM_018194.5 NP_060664.2 8 - 9 1249 93
XM_006711441.3 XP_006711504.1 9 - 10 1310 93
XM_011509737.2 XP_011508039.1 9 - 10 1310 93
XM_011509738.2 XP_011508040.1 7 - 8 1068 93
XM_011509739.2 XP_011508041.1 8 - 9 1235 93
XM_011509740.2 XP_011508042.1 9 - 10 1439 93
XM_011509741.2 XP_011508043.1 8 - 9 1183 93
XM_011509742.2 XP_011508044.1 8 - 9 1092 93
XM_011509744.2 XP_011508046.1 7 - 8 1243 93
XM_011509746.2 XP_011508048.1 7 - 8 1069 93
XM_011509747.1 XP_011508049.1 6 - 7 1033 93
XM_017001726.1 XP_016857215.1 7 - 8 1284 93
XM_017001727.1 XP_016857216.1 7 - 8 1284 93
XM_017001728.1 XP_016857217.1 8 - 9 1393 93
XM_017001729.1 XP_016857218.1 8 - 9 1114 93
XM_017001730.1 XP_016857219.1 7 - 8 1196 93
XM_017001731.1 XP_016857220.1 7 - 8 1002 93
XM_017001732.1 XP_016857221.1 7 - 8 879 93
XM_017001733.1 XP_016857222.1 6 - 7 1156 93
XM_017001734.1 XP_016857223.1 7 - 8 1190 93
XM_017001735.1 XP_016857224.1 7 - 8 1519 93
XM_017001736.1 XP_016857225.1 7 - 8 1090 93
XM_017001737.1 XP_016857226.1 6 - 7 663 93
XM_017001738.1 XP_016857227.1 6 - 7 770 93
XM_017001739.1 XP_016857228.1 6 - 7 903 93
XM_017001740.1 XP_016857229.1 6 - 7 1404 93
XM_017001741.1 XP_016857230.1 4 - 5 565 93
XM_017001742.1 XP_016857231.1 4 - 5 1204 93
XM_017001743.1 XP_016857232.1 4 - 5 561 93
XM_017001744.1 XP_016857233.1 4 - 5 993 93
GenBank mRNA AK001586.1 - 8 - 9 1203 93
AK092186.1 - 2 - 3 322 93
AK295130.1 - 7 - 8 1135 93
AK297193.1 - 6 - 7 770 93
AK298991.1 - 7 - 8 1008 93
AK299200.1 - 6 - 7 1033 93
AK302955.1 - 7 - 8 1068 93
AK316190.1 - 7 - 8 1069 93
AK316524.1 - 7 - 8 1101 93
BC039071.1 - 8 - 9 1188 93
BC117130.1 - 7 - 8 1038 93

View Full Product Details