Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEP15
Entrez Gene ID
9403
Gene Name
15 kDa selenoprotein
Gene Aliases
-
Chromosome Location
Chr.1: 86862445 - 86914424 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203341.1 NP_976086.1 3 - 4 629 88
GenBank mRNA BC021697.1 - 3 - 4 343 88

View Full Product Details