The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321786.1 | NP_001308715.1 | 16 - 17 | 2655 | 67 |
NM_001321788.1 | NP_001308717.1 | 16 - 17 | 2574 | 67 | |
NM_001321789.1 | NP_001308718.1 | 15 - 16 | 2446 | 67 | |
NM_001321794.1 | NP_001308723.1 | 15 - 16 | 2427 | 67 | |
NM_001321795.1 | NP_001308724.1 | 15 - 16 | 2603 | 67 | |
NM_001321796.1 | NP_001308725.1 | 15 - 16 | 2427 | 67 | |
NM_001321806.1 | NP_001308735.1 | 15 - 16 | 2417 | 67 | |
NM_001321807.1 | NP_001308736.1 | 17 - 18 | 2894 | 67 | |
NM_001321811.1 | NP_001308740.1 | 14 - 15 | 2446 | 67 | |
NM_001321813.1 | NP_001308742.1 | 16 - 17 | 2484 | 67 | |
NM_001321822.1 | NP_001308751.1 | 17 - 18 | 2800 | 67 | |
NM_170662.4 | NP_733762.2 | 16 - 17 | 2750 | 67 | |
NR_135806.1 | 17 - 18 | 2882 | 67 | ||
NR_135807.1 | 17 - 18 | 2798 | 67 | ||
NR_135808.1 | 16 - 17 | 2750 | 67 | ||
NR_135810.1 | 16 - 17 | 2528 | 67 | ||
NR_135811.1 | 16 - 17 | 2574 | 67 | ||
XM_011513257.1 | XP_011511559.1 | 16 - 17 | 2570 | 67 | |
XM_011513259.2 | XP_011511561.2 | 16 - 17 | 2695 | 67 | |
XM_017007395.1 | XP_016862884.1 | 16 - 17 | 2522 | 67 | |
XM_017007397.1 | XP_016862886.1 | 15 - 16 | 2547 | 67 | |
XM_017007398.1 | XP_016862887.1 | 15 - 16 | 2373 | 67 | |
XM_017007399.1 | XP_016862888.1 | 15 - 16 | 2598 | 67 | |
GenBank mRNA | AB075490.1 | - | 5 - 6 | 1033 | 67 |
AK094184.1 | - | 7 - 8 | 976 | 67 | |
AK292792.1 | - | 16 - 17 | 2691 | 67 | |
BC032851.2 | - | 16 - 17 | 2691 | 67 | |
DQ349203.1 | - | 15 - 16 | 2428 | 67 | |
DQ349204.1 | - | 15 - 16 | 2428 | 67 | |
U26710.1 | - | 16 - 17 | 2750 | 67 |