Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ORC4
Entrez Gene ID
5000
Gene Name
origin recognition complex subunit 4
Gene Aliases
ORC4L, ORC4P
Chromosome Location
Chr.2: 147930397 - 148021604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017004226.1 XP_016859715.1 2 - 3 312 92

View Full Product Details