The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001181.4 | NP_001172.1 | 5 - 6 | 686 | 108 |
NM_001201352.1 | NP_001188281.1 | 5 - 6 | 671 | 108 | |
NM_080912.3 | NP_550434.1 | 5 - 6 | 774 | 108 | |
NM_080913.3 | NP_550435.1 | 5 - 6 | 614 | 108 | |
NM_080914.2 | NP_550436.1 | 4 - 5 | 432 | 108 | |
XM_005256648.2 | XP_005256705.1 | 6 - 7 | 724 | 108 | |
XM_006721524.2 | XP_006721587.2 | 4 - 5 | 491 | 108 | |
XM_006721526.1 | XP_006721589.1 | 6 - 7 | 1171 | 108 | |
XM_011523862.2 | XP_011522164.2 | 6 - 7 | 777 | 108 | |
XM_011523863.2 | XP_011522165.1 | 5 - 6 | 646 | 108 | |
XM_011523864.2 | XP_011522166.1 | 5 - 6 | 681 | 108 | |
XM_011523865.1 | XP_011522167.1 | 6 - 7 | 1207 | 108 | |
XM_011523866.1 | XP_011522168.1 | 6 - 7 | 757 | 108 | |
XM_017024651.1 | XP_016880140.1 | 5 - 6 | 625 | 108 | |
XM_017024652.1 | XP_016880141.1 | 6 - 7 | 1155 | 108 | |
XM_017024653.1 | XP_016880142.1 | 6 - 7 | 1243 | 108 | |
GenBank mRNA | AF529374.1 | - | 4 - 5 | 404 | 108 |
BC017251.2 | - | 5 - 6 | 562 | 108 | |
M11025.1 | - | 5 - 6 | 539 | 108 | |
U97197.1 | - | 4 - 5 | 362 | 108 | |
X55283.1 | - | 5 - 6 | 537 | 108 | |
X55284.1 | - | 5 - 6 | 616 | 108 |