Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASGR2
Entrez Gene ID
433
Gene Name
asialoglycoprotein receptor 2
Gene Aliases
ASGP-R2, ASGPR2, CLEC4H2, HBXBP, HL-2
Chromosome Location
Chr.17: 7101322 - 7115522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080913.3 NP_550435.1 2 - 3 341 85
NM_080914.2 NP_550436.1 1 - 2 144 85
XM_005256648.2 XP_005256705.1 3 - 4 451 85
XM_006721526.1 XP_006721589.1 3 - 4 898 85
XM_011523863.2 XP_011522165.1 2 - 3 337 85
XM_011523865.1 XP_011522167.1 3 - 4 898 85
XM_011523866.1 XP_011522168.1 3 - 4 448 85
XM_017024651.1 XP_016880140.1 2 - 3 337 85
GenBank mRNA AK303787.1 - 3 - 4 457 85
AK310123.1 - 2 - 3 443 85
BC017251.2 - 2 - 3 289 85
BX483468.1 - 2 - 3 354 85
DC315727.1 - 3 - 4 453 85
U97197.1 - 1 - 2 74 85
X55283.1 - 2 - 3 264 85

View Full Product Details