The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000950.2 | NP_000941.1 | 2 - 3 | 213 | 100 |
NM_001173489.1 | NP_001166960.1 | 2 - 3 | 232 | 100 | |
GenBank mRNA | AF009242.1 | - | 2 - 3 | 125 | 100 |
DA245329.1 | - | 2 - 3 | 199 | 100 | |
DA411839.1 | - | 2 - 3 | 171 | 100 |