The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122853.2 | NP_001116325.1 | 7 - 7 | 107 | |
NM_133371.4 | NP_588612.2 | 7 - 7 | 107 | ||
GenBank mRNA | AA969304.1 | - | 1 - 1 | 112 | 107 |
AF052497.1 | - | 2 - 2 | 107 | ||
AJ300586.1 | - | 7 - 7 | 107 |