Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJA3
Entrez Gene ID
9093
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
Gene Aliases
HCA57, TID1, hTID-1
Chromosome Location
Chr.16: 4425805 - 4456775 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005147.5 NP_005138.3 11 - 12 1532 130
GenBank mRNA AB209923.1 - 11 - 12 1451 130
AF061749.1 - 11 - 12 1486 130
AK127660.1 - 2 - 3 1060 130
AK223105.1 - 11 - 12 1472 130
BC007225.1 - 11 - 12 1452 130
BC011855.2 - 11 - 12 1456 130

View Full Product Details