Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HHAT
Entrez Gene ID
55733
Gene Name
hedgehog acyltransferase
Gene Aliases
MART2, SKI1, Skn
Chromosome Location
Chr.1: 210327873 - 210676298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170588.2 NP_001164059.1 1 - 2 202 90
NM_018194.5 NP_060664.2 1 - 2 202 90
XM_011509739.2 XP_011508041.1 1 - 2 188 90
XM_011509746.2 XP_011508048.1 1 - 2 217 90
XM_017001730.1 XP_016857219.1 1 - 2 217 90
XM_017001734.1 XP_016857223.1 1 - 2 211 90
XM_017001739.1 XP_016857228.1 1 - 2 223 90
GenBank mRNA AK001586.1 - 1 - 2 156 90
AK295130.1 - 1 - 2 156 90
AK298991.1 - 1 - 2 156 90
BC039071.1 - 1 - 2 144 90
DA336461.1 - 1 - 2 188 90

View Full Product Details