Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PARP1
Entrez Gene ID
142
Gene Name
poly(ADP-ribose) polymerase 1
Gene Aliases
ADPRT, ADPRT 1, ADPRT1, ARTD1, PARP, PARP-1, PPOL, pADPRT-1
Chromosome Location
Chr.1: 226360691 - 226408100 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001618.3 NP_001609.2 21 - 22 3018 140
GenBank mRNA AK125650.1 - 6 - 7 2802 140
AK225654.1 - 21 - 22 2983 140
AK303340.1 - 21 - 22 2945 140
AK312339.1 - 21 - 22 2934 140
BC037545.1 - 21 - 22 2991 140
J03473.1 - 21 - 22 2942 140
M17081.1 - 12 - 13 1527 140
M18112.1 - 21 - 22 2986 140
M32721.1 - 21 - 22 3006 140

View Full Product Details