Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PARP1
Entrez Gene ID
142
Gene Name
poly(ADP-ribose) polymerase 1
Gene Aliases
ADPRT, ADPRT 1, ADPRT1, ARTD1, PARP, PARP-1, PPOL, pADPRT-1
Chromosome Location
Chr.1: 226360691 - 226408100 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001618.3 NP_001609.2 5 - 6 888 107
GenBank mRNA AK225654.1 - 5 - 6 853 107
AK303340.1 - 5 - 6 815 107
AK312339.1 - 5 - 6 804 107
BC037545.1 - 5 - 6 861 107
J03473.1 - 5 - 6 812 107
M18112.1 - 5 - 6 856 107
M32721.1 - 5 - 6 876 107

View Full Product Details