Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC24D
Entrez Gene ID
9871
Gene Name
SEC24 homolog D, COPII coat complex component
Gene Aliases
CLCRP2
Chromosome Location
Chr.4: 118722823 - 118838683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005263379.2 XP_005263436.1 1 - 2 125 128
GenBank mRNA AK310259.1 - 1 - 2 114 128
BC037736.1 - 1 - 2 117 128

View Full Product Details