The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168320.1 | NP_001161792.1 | 5 - 6 | 554 | 69 |
NM_006675.4 | NP_006666.1 | 6 - 7 | 621 | 69 | |
XM_011520912.2 | XP_011519214.1 | 7 - 8 | 738 | 69 | |
XM_011520913.2 | XP_011519215.1 | 4 - 5 | 480 | 69 | |
GenBank mRNA | AF089749.1 | - | 6 - 7 | 564 | 69 |
AK027366.1 | - | 6 - 7 | 597 | 69 | |
AK223179.1 | - | 6 - 7 | 593 | 69 | |
BC071881.1 | - | 5 - 6 | 536 | 69 | |
CR542109.1 | - | 4 - 5 | 438 | 69 |