The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031942.4 | NP_114148.3 | 4 - 5 | 758 | 149 |
NM_145810.2 | NP_665809.1 | 3 - 4 | 521 | 149 | |
XM_011511957.2 | XP_011510259.1 | 4 - 5 | 1299 | 149 | |
GenBank mRNA | AK027628.1 | - | 4 - 5 | 730 | 149 |
AK027642.1 | - | 3 - 4 | 502 | 149 | |
AK075134.1 | - | 3 - 4 | 482 | 149 | |
AK223524.1 | - | 3 - 4 | 526 | 149 | |
AK297097.1 | - | 2 - 3 | 356 | 149 | |
AK297245.1 | - | 4 - 5 | 618 | 149 | |
AK300949.1 | - | 4 - 5 | 587 | 149 | |
AL834186.1 | - | 3 - 4 | 461 | 149 | |
AY029179.1 | - | 3 - 4 | 534 | 149 | |
BC027966.1 | - | 3 - 4 | 487 | 149 |