Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDCA7
Entrez Gene ID
83879
Gene Name
cell division cycle associated 7
Gene Aliases
ICF3, JPO1
Chromosome Location
Chr.2: 173354017 - 173368992 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031942.4 NP_114148.3 9 - 10 1455 81
NM_145810.2 NP_665809.1 8 - 9 1218 81
XM_011511957.2 XP_011510259.1 9 - 10 1996 81
GenBank mRNA AK027628.1 - 9 - 10 1427 81
AK027642.1 - 8 - 9 1199 81
AK075134.1 - 7 - 8 2081 81
AK297097.1 - 7 - 8 1053 81
AK297245.1 - 9 - 10 1315 81
AK300949.1 - 9 - 10 1284 81
AL833728.1 - 4 - 5 747 81
AL834186.1 - 6 - 7 1909 81
AY029179.1 - 8 - 9 1231 81
BC015124.1 - 4 - 5 634 81
BC027966.1 - 8 - 9 1184 81

View Full Product Details