Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTPAP
Entrez Gene ID
55149
Gene Name
mitochondrial poly(A) polymerase
Gene Aliases
PAPD1, SPAX4
Chromosome Location
Chr.10: 30309801 - 30349338 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018109.3 NP_060579.3 3 - 4 625 94
GenBank mRNA AB194709.1 - 3 - 4 562 94
AK001348.1 - 3 - 4 566 94
AK022188.1 - 3 - 4 605 94
AY364242.1 - 3 - 4 562 94
BC061703.1 - 3 - 4 620 94

View Full Product Details