The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000755.3 | NP_000746.2 | 5 - 6 | 909 | 65 |
NM_001257363.1 | NP_001244292.1 | 6 - 7 | 1008 | 65 | |
XM_005251708.3 | XP_005251765.1 | 7 - 8 | 1071 | 65 | |
XM_017014274.1 | XP_016869763.1 | 6 - 7 | 1076 | 65 | |
XM_017014275.1 | XP_016869764.1 | 5 - 6 | 729 | 65 | |
GenBank mRNA | AK313931.1 | - | 6 - 7 | 920 | 65 |
BC000723.2 | - | 5 - 6 | 848 | 65 | |
BT006801.1 | - | 5 - 6 | 627 | 65 | |
X78706.1 | - | 5 - 6 | 622 | 65 |