The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134670.1 | NP_001128142.1 | 1 - 2 | 572 | 137 |
GenBank mRNA | BC011916.2 | - | 1 - 2 | 572 | 137 |
BC057821.1 | - | 1 - 2 | 546 | 137 |