Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALDH18A1
Entrez Gene ID
5832
Gene Name
aldehyde dehydrogenase 18 family member A1
Gene Aliases
ADCL3, ARCL3A, GSAS, P5CS, PYCS, SPG9A, SPG9B
Chromosome Location
Chr.10: 95605929 - 95656810 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017423.1 NP_001017423.1 13 - 14 1840 75
NM_001323412.1 NP_001310341.1 12 - 13 1631 75
NM_001323413.1 NP_001310342.1 13 - 14 1997 75
NM_001323414.1 NP_001310343.1 13 - 14 1866 75
NM_001323415.1 NP_001310344.1 13 - 14 1991 75
NM_001323416.1 NP_001310345.1 12 - 13 1782 75
NM_001323417.1 NP_001310346.1 12 - 13 1741 75
NM_001323418.1 NP_001310347.1 12 - 13 1776 75
NM_001323419.1 NP_001310348.1 10 - 11 1376 75
NM_002860.3 NP_002851.2 13 - 14 1846 75
GenBank mRNA AK225455.1 - 12 - 13 1416 75
AK225465.1 - 12 - 13 1416 75
AK226032.1 - 12 - 13 1416 75
AK295487.1 - 12 - 13 1497 75
AK299557.1 - 10 - 11 1274 75
AK312271.1 - 13 - 14 1748 75
BC106054.1 - 13 - 14 1681 75
BC117240.1 - 13 - 14 1787 75
BC117242.1 - 13 - 14 1787 75
BC143930.1 - 13 - 14 1781 75
U68758.1 - 13 - 14 1655 75
U76542.1 - 13 - 14 1661 75
X94453.1 - 13 - 14 1742 75

View Full Product Details