The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005558.3 | NP_005549.2 | 3 - 4 | 1267 | 124 |
GenBank mRNA | AK310917.1 | - | 4 - 5 | 1153 | 124 |
AK313388.1 | - | 3 - 4 | 1050 | 124 | |
BC009742.2 | - | 3 - 4 | 1267 | 124 | |
BC071890.1 | - | 3 - 4 | 1328 | 124 | |
BX647307.1 | - | 3 - 4 | 1072 | 124 | |
U42408.1 | - | 3 - 4 | 1251 | 124 |