The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288563.1 | NP_001275492.1 | 2 - 3 | 266 | 108 |
NM_145172.4 | NP_660155.2 | 2 - 3 | 266 | 108 | |
GenBank mRNA | AK054629.1 | - | 2 - 3 | 220 | 108 |
AK292603.1 | - | 2 - 3 | 237 | 108 | |
AY049724.1 | - | 2 - 3 | 247 | 108 | |
BC040265.1 | - | 2 - 3 | 220 | 108 | |
BX648851.1 | - | 2 - 3 | 270 | 108 | |
DB075669.1 | - | 2 - 3 | 243 | 108 | |
DB085334.1 | - | 2 - 3 | 266 | 108 |