The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016578.3 | NP_057662.3 | 6 - 7 | 2633 | 118 |
XM_017017923.1 | XP_016873412.1 | 6 - 7 | 2407 | 118 | |
GenBank mRNA | AF227948.2 | - | 1 - 2 | 2818 | 118 |
AF380176.1 | - | 6 - 7 | 2504 | 118 | |
AK001268.1 | - | 1 - 2 | 1055 | 118 | |
BC046124.1 | - | 2 - 3 | 1916 | 118 |