The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178536.3 | NP_848631.2 | 4 - 5 | 464 | 80 |
XM_006717065.3 | XP_006717128.1 | 5 - 6 | 949 | 80 | |
XM_011518560.2 | XP_011516862.1 | 5 - 6 | 1101 | 80 | |
XM_011518561.2 | XP_011516863.2 | 4 - 5 | 641 | 80 | |
XM_011518562.2 | XP_011516864.1 | 5 - 6 | 502 | 80 | |
XM_017014631.1 | XP_016870120.1 | 4 - 5 | 640 | 80 | |
XM_017014632.1 | XP_016870121.1 | 4 - 5 | 560 | 80 | |
GenBank mRNA | AI681012.1 | - | 2 - 3 | 80 | |
AY301272.1 | - | 4 - 5 | 452 | 80 | |
BC041168.1 | - | 4 - 5 | 464 | 80 |