Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC24D
Entrez Gene ID
9871
Gene Name
SEC24 homolog D, COPII coat complex component
Gene Aliases
CLCRP2
Chromosome Location
Chr.4: 118722823 - 118838683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318066.1 NP_001304995.1 18 - 19 2677 128
NM_014822.3 NP_055637.2 18 - 19 2674 128
XM_005263379.2 XP_005263436.1 18 - 19 2551 128
XM_017008875.1 XP_016864364.1 13 - 14 1832 128
GenBank mRNA AB018298.1 - 18 - 19 2577 128
AF130464.2 - 17 - 18 2377 128
AK001390.1 - 15 - 16 2045 128
AK125962.1 - 12 - 13 1665 128
AK291765.1 - 18 - 19 2574 128
AK309960.1 - 11 - 12 1413 128
BC035761.1 - 18 - 19 2599 128
BC037736.1 - 21 - 22 3151 128

View Full Product Details