Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LTF
Entrez Gene ID
4057
Gene Name
lactotransferrin
Gene Aliases
GIG12, HEL110, HLF2, LF
Chromosome Location
Chr.3: 46436005 - 46485234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199149.1 NP_001186078.1 13 - 14 1839 87
NM_001321121.1 NP_001308050.1 13 - 14 1944 87
NM_001321122.1 NP_001308051.1 16 - 17 2183 87
NM_002343.5 NP_002334.2 13 - 14 1950 87
GenBank mRNA AF332168.1 - 13 - 14 1657 87
AK093852.1 - 16 - 17 2186 87
AK290859.1 - 13 - 14 1695 87
AK292813.1 - 13 - 14 1695 87
AK298035.1 - 13 - 14 1839 87
AK303995.1 - 5 - 6 558 87
AK316330.1 - 13 - 14 1759 87
AY137470.1 - 13 - 14 1657 87
AY165046.1 - 13 - 14 1669 87
AY178998.2 - 13 - 14 1657 87
AY360320.1 - 13 - 14 1652 87
AY493417.1 - 13 - 14 1767 87
BC015823.1 - 13 - 14 1701 87
BC022347.1 - 13 - 14 1695 87
EU794669.1 - 13 - 14 1692 87
KF992208.1 - 13 - 14 1669 87
KT006756.1 - 13 - 14 1657 87
M18642.1 - 3 - 4 352 87
M83202.1 - 13 - 14 1664 87
M83205.1 - 13 - 14 1643 87
M93150.1 - 13 - 14 1674 87
U07643.1 - 13 - 14 1754 87
X52941.1 - 13 - 14 1651 87
X53961.1 - 13 - 14 1951 87

View Full Product Details