Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LTF
Entrez Gene ID
4057
Gene Name
lactotransferrin
Gene Aliases
GIG12, HEL110, HLF2, LF
Chromosome Location
Chr.3: 46436005 - 46485234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199149.1 NP_001186078.1 4 - 5 684 56
NM_001321121.1 NP_001308050.1 4 - 5 795 56
NM_001321122.1 NP_001308051.1 7 - 8 1028 56
NM_002343.5 NP_002334.2 4 - 5 795 56
GenBank mRNA AF332168.1 - 4 - 5 502 56
AK093852.1 - 7 - 8 1031 56
AK290859.1 - 4 - 5 540 56
AK292813.1 - 4 - 5 540 56
AK298035.1 - 4 - 5 684 56
AK316330.1 - 4 - 5 604 56
AY137470.1 - 4 - 5 502 56
AY165046.1 - 4 - 5 514 56
AY178998.2 - 4 - 5 502 56
AY360320.1 - 4 - 5 503 56
AY493417.1 - 4 - 5 612 56
AY875691.1 - 4 - 5 502 56
BC015822.1 - 4 - 5 544 56
BC015823.1 - 4 - 5 546 56
BC022347.1 - 4 - 5 540 56
CD722125.1 - 4 - 5 528 56
DQ522304.1 - 4 - 5 502 56
EU794669.1 - 4 - 5 537 56
KF992208.1 - 4 - 5 514 56
KT006756.1 - 4 - 5 502 56
M83202.1 - 4 - 5 509 56
M83205.1 - 4 - 5 488 56
M93150.1 - 4 - 5 519 56
U07643.1 - 4 - 5 599 56
X52941.1 - 4 - 5 496 56
X53961.1 - 4 - 5 796 56

View Full Product Details