The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146008.1 | NP_001139480.1 | 1 - 2 | 1629 | 96 |
NM_001146009.1 | NP_001139481.1 | 1 - 2 | 1408 | 96 | |
NM_030958.2 | NP_112220.2 | 2 - 3 | 1619 | 96 | |
XM_005251313.2 | XP_005251370.1 | 2 - 3 | 1619 | 96 | |
XM_017013883.1 | XP_016869372.1 | 2 - 3 | 1619 | 96 | |
XM_017013884.1 | XP_016869373.1 | 2 - 3 | 1619 | 96 | |
XM_017013886.1 | XP_016869375.1 | 2 - 3 | 1619 | 96 | |
GenBank mRNA | AF205075.1 | - | 2 - 3 | 1528 | 96 |
AK022760.1 | - | 1 - 2 | 1629 | 96 | |
AK096879.1 | - | 2 - 3 | 1619 | 96 | |
AK299048.1 | - | 2 - 3 | 1575 | 96 | |
AK307567.1 | - | 2 - 3 | 1577 | 96 | |
AK308488.1 | - | 2 - 3 | 1521 | 96 | |
BC137424.1 | - | 1 - 2 | 1414 | 96 | |
BC139755.1 | - | 1 - 2 | 954 | 96 |