The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146009.1 | NP_001139481.1 | 6 - 7 | 2352 | 64 |
NM_030958.2 | NP_112220.2 | 8 - 9 | 2728 | 64 | |
XM_005251313.2 | XP_005251370.1 | 7 - 8 | 2563 | 64 | |
XM_017013885.1 | XP_016869374.1 | 7 - 8 | 2371 | 64 | |
GenBank mRNA | AF205075.1 | - | 8 - 9 | 2637 | 64 |
BC137424.1 | - | 7 - 8 | 2523 | 64 | |
BC139755.1 | - | 6 - 7 | 1898 | 64 | |
BC144618.1 | - | 3 - 4 | 1808 | 64 |