The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146009.1 | NP_001139481.1 | 7 - 8 | 2414 | 106 |
NM_030958.2 | NP_112220.2 | 9 - 10 | 2790 | 106 | |
XM_005251313.2 | XP_005251370.1 | 8 - 9 | 2625 | 106 | |
XM_017013885.1 | XP_016869374.1 | 8 - 9 | 2433 | 106 | |
GenBank mRNA | AF205075.1 | - | 9 - 10 | 2699 | 106 |
BC137424.1 | - | 8 - 9 | 2585 | 106 | |
BC139755.1 | - | 7 - 8 | 1960 | 106 | |
BC144618.1 | - | 4 - 5 | 1870 | 106 |