The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022479.2 | NP_071924.1 | 10 - 11 | 1931 | 68 |
GenBank mRNA | AB078148.1 | - | 10 - 11 | 1665 | 68 |
AF410457.1 | - | 10 - 11 | 1665 | 68 | |
AJ626726.1 | - | 10 - 11 | 1665 | 68 | |
AK022143.1 | - | 2 - 3 | 199 | 68 | |
AK091379.1 | - | 6 - 7 | 881 | 68 | |
AK094958.1 | - | 10 - 11 | 1599 | 68 | |
AK126044.1 | - | 10 - 11 | 2067 | 68 | |
AK290036.1 | - | 10 - 11 | 1934 | 68 | |
AL137431.1 | - | 2 - 3 | 272 | 68 | |
BC067524.1 | - | 10 - 11 | 1665 | 68 | |
BC067525.1 | - | 10 - 11 | 1665 | 68 | |
BC069624.1 | - | 10 - 11 | 1665 | 68 | |
BC069628.1 | - | 10 - 11 | 1665 | 68 | |
BC069636.1 | - | 10 - 11 | 1665 | 68 | |
BC069645.1 | - | 10 - 11 | 1665 | 68 | |
BC069997.1 | - | 10 - 11 | 1665 | 68 |