The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022479.2 | NP_071924.1 | 8 - 9 | 1670 | 154 |
XM_011516467.2 | XP_011514769.1 | 8 - 9 | 1793 | 154 | |
GenBank mRNA | AB078148.1 | - | 8 - 9 | 1404 | 154 |
AF410457.1 | - | 8 - 9 | 1404 | 154 | |
AJ626726.1 | - | 8 - 9 | 1404 | 154 | |
AK091379.1 | - | 4 - 5 | 620 | 154 | |
AK094958.1 | - | 8 - 9 | 1338 | 154 | |
AK126044.1 | - | 8 - 9 | 1806 | 154 | |
AK290036.1 | - | 8 - 9 | 1673 | 154 | |
BC067524.1 | - | 8 - 9 | 1404 | 154 | |
BC067525.1 | - | 8 - 9 | 1404 | 154 | |
BC069624.1 | - | 8 - 9 | 1404 | 154 | |
BC069628.1 | - | 8 - 9 | 1404 | 154 | |
BC069636.1 | - | 8 - 9 | 1404 | 154 | |
BC069645.1 | - | 8 - 9 | 1404 | 154 | |
BC069997.1 | - | 8 - 9 | 1404 | 154 |