The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002819.4 | NP_002810.1 | 10 - 11 | 1226 | 68 |
NM_031990.3 | NP_114367.1 | 10 - 11 | 1205 | 68 | |
NM_031991.3 | NP_114368.1 | 9 - 10 | 1148 | 68 | |
XM_005259597.2 | XP_005259654.1 | 10 - 11 | 1729 | 68 | |
XM_005259598.2 | XP_005259655.1 | 9 - 10 | 1651 | 68 | |
GenBank mRNA | BC002397.2 | - | 10 - 11 | 1193 | 68 |
BC004383.1 | - | 9 - 10 | 1058 | 68 | |
BC013694.1 | - | 10 - 11 | 1139 | 68 | |
BC023219.1 | - | 10 - 11 | 1192 | 68 | |
BT006819.1 | - | 9 - 10 | 1120 | 68 | |
X60648.1 | - | 9 - 10 | 1070 | 68 | |
X62006.1 | - | 9 - 10 | 1087 | 68 | |
X65371.1 | - | 10 - 11 | 1144 | 68 | |
X65372.1 | - | 10 - 11 | 1165 | 68 | |
X66975.1 | - | 10 - 11 | 1207 | 68 |