Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIFM1
Entrez Gene ID
9131
Gene Name
apoptosis inducing factor, mitochondria associated 1
Gene Aliases
AIF, CMT2D, CMTX4, COWCK, COXPD6, DFNX5, NADMR, NAMSD, PDCD8
Chromosome Location
Chr.X: 130129362 - 130165887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004208.3 NP_004199.1 9 - 10 1191 132
NM_145812.2 NP_665811.1 9 - 10 1179 132
GenBank mRNA AF100928.1 - 9 - 10 1002 132
AF131759.1 - 6 - 7 543 132
AK000775.1 - 9 - 10 1027 132
AK314446.1 - 9 - 10 979 132
AL049703.1 - 9 - 10 1044 132
BC111065.1 - 9 - 10 1122 132
BC139738.1 - 9 - 10 1095 132
CR457379.1 - 9 - 10 960 132

View Full Product Details