The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021733.1 | NP_068379.1 | 10 - 11 | 1627 | 74 |
XM_011527202.1 | XP_011525504.1 | 10 - 11 | 1740 | 74 | |
GenBank mRNA | AF411384.1 | - | 6 - 7 | 1166 | 74 |
BC058862.1 | - | 10 - 11 | 1655 | 74 |