The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021733.1 | NP_068379.1 | 7 - 8 | 1200 | 65 |
XM_011527201.1 | XP_011525503.1 | 7 - 8 | 1270 | 65 | |
XM_011527202.1 | XP_011525504.1 | 7 - 8 | 1270 | 65 | |
XM_011527203.2 | XP_011525505.1 | 7 - 8 | 1270 | 65 | |
GenBank mRNA | AF411384.1 | - | 3 - 4 | 739 | 65 |
BC058862.1 | - | 7 - 8 | 1228 | 65 |