The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021733.1 | NP_068379.1 | 9 - 10 | 1512 | 67 |
XM_011527203.2 | XP_011525505.1 | 9 - 10 | 1582 | 67 | |
GenBank mRNA | AF411384.1 | - | 5 - 6 | 1051 | 67 |
BC058862.1 | - | 9 - 10 | 1540 | 67 |