Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTHFD1L
Entrez Gene ID
25902
Gene Name
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like
Gene Aliases
FTHFSDC1, MTC1THFS, dJ292B18.2
Chromosome Location
Chr.6: 150865679 - 151101887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242767.1 NP_001229696.1 3 - 4 506 74
NM_001242768.1 NP_001229697.1 3 - 4 190 74
NM_001242769.1 NP_001229698.1 3 - 4 506 74
NM_015440.4 NP_056255.2 3 - 4 506 74
XM_005266907.4 XP_005266964.1 3 - 4 447 74
XM_005266910.4 XP_005266967.1 3 - 4 447 74
XM_005266911.4 XP_005266968.1 3 - 4 447 74
XM_011535729.2 XP_011534031.1 3 - 4 447 74
XM_011535730.2 XP_011534032.1 3 - 4 190 74
XM_011535731.2 XP_011534033.1 3 - 4 165 74
XM_011535732.2 XP_011534034.1 3 - 4 244 74
XM_011535733.2 XP_011534035.1 3 - 4 690 74
XM_011535734.2 XP_011534036.1 3 - 4 286 74
XM_011535737.2 XP_011534039.1 3 - 4 447 74
XM_011535738.2 XP_011534040.1 3 - 4 447 74
XM_017010702.1 XP_016866191.1 3 - 4 446 74
XM_017010703.1 XP_016866192.1 3 - 4 446 74
XM_017010704.1 XP_016866193.1 3 - 4 165 74
XM_017010705.1 XP_016866194.1 3 - 4 690 74
XM_017010706.1 XP_016866195.1 3 - 4 690 74
XM_017010707.1 XP_016866196.1 3 - 4 690 74
XM_017010708.1 XP_016866197.1 3 - 4 448 74
GenBank mRNA AB127387.1 - 3 - 4 362 74
AK127089.1 - 2 - 3 982 74
AK310342.1 - 3 - 4 1223 74
AK315378.1 - 3 - 4 190 74
AL117452.1 - 3 - 4 181 74
AV704883.1 - 3 - 4 241 74
AY374130.1 - 3 - 4 469 74
AY374131.1 - 3 - 4 469 74
BC110319.1 - 2 - 3 128 74
BC136685.1 - 3 - 4 396 74
BC144368.1 - 3 - 4 396 74
BC144369.1 - 3 - 4 396 74
BQ062382.1 - - 488 74
DA195967.1 - 3 - 4 190 74

View Full Product Details