The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135691.2 | NP_001129163.1 | 14 - 15 | 1686 | 121 |
NM_001142324.1 | NP_001135796.1 | 13 - 14 | 1590 | 121 | |
NM_001142325.1 | NP_001135797.1 | 14 - 15 | 1686 | 121 | |
NM_003053.3 | NP_003044.1 | 13 - 14 | 1482 | 121 | |
XM_011544625.1 | XP_011542927.1 | 12 - 13 | 1331 | 121 | |
XM_011544626.1 | XP_011542928.1 | 12 - 13 | 1319 | 121 | |
XM_011544627.1 | XP_011542929.1 | 13 - 14 | 1415 | 121 | |
GenBank mRNA | AK313720.1 | - | 13 - 14 | 1378 | 121 |
BC006317.2 | - | 13 - 14 | 1499 | 121 | |
U39905.1 | - | 13 - 14 | 1482 | 121 |