Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX7
Entrez Gene ID
5191
Gene Name
peroxisomal biogenesis factor 7
Gene Aliases
PBD9B, PTS2R, RCDP1, RD
Chromosome Location
Chr.6: 136822220 - 136913934 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000288.3 NP_000279.1 3 - 4 437 64
XM_005267019.4 XP_005267076.1 3 - 4 717 64
XM_017010934.1 XP_016866423.1 3 - 4 781 64
GenBank mRNA BC006268.1 - 3 - 4 379 64
BC031606.1 - 3 - 4 421 64
BP246238.1 - 3 - 4 437 64
CR542054.1 - 3 - 4 335 64
CR542076.1 - 3 - 4 335 64
U76560.1 - 3 - 4 383 64
U88871.1 - 3 - 4 416 64

View Full Product Details