The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130915.1 | NP_001124387.1 | 1 - 2 | 40 | 68 |
XM_011526808.2 | XP_011525110.1 | 1 - 2 | 219 | 68 | |
GenBank mRNA | AK131427.1 | - | 1 - 2 | 47 | 68 |