The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270550.1 | NP_001257479.1 | 1 - 2 | 403 | 124 |
GenBank mRNA | AK308210.1 | - | 1 - 2 | 276 | 124 |
AK309963.1 | - | 1 - 2 | 403 | 124 | |
CN266369.1 | - | 1 - 2 | 99 | 124 |