Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MBD1
Entrez Gene ID
4152
Gene Name
methyl-CpG binding domain protein 1
Gene Aliases
CXXC3, PCM1, RFT
Chromosome Location
Chr.18: 50266882 - 50281774 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204136.1 NP_001191065.1 13 - 14 2025 82
NM_001204137.1 NP_001191066.1 14 - 15 2100 82
NM_001204138.1 NP_001191067.1 14 - 15 2097 82
NM_001204139.1 NP_001191068.1 13 - 14 2025 82
NM_001204140.1 NP_001191069.1 13 - 14 1932 82
NM_001204141.1 NP_001191070.1 12 - 13 1875 82
NM_001323942.1 NP_001310871.1 14 - 15 2100 82
NM_001323947.1 NP_001310876.1 14 - 15 2055 82
NM_001323949.1 NP_001310878.1 14 - 15 2092 82
NM_001323950.1 NP_001310879.1 13 - 14 1977 82
NM_001323951.1 NP_001310880.1 13 - 14 1980 82
NM_001323952.1 NP_001310881.1 12 - 13 1782 82
NM_001323954.1 NP_001310883.1 11 - 12 1788 82
NM_015844.2 NP_056669.2 12 - 13 1857 82
NM_015846.3 NP_056671.2 13 - 14 2025 82
NM_015847.3 NP_056723.2 12 - 13 1878 82
XM_005258271.2 XP_005258328.1 13 - 14 1772 82
XM_006722456.2 XP_006722519.1 14 - 15 1847 82
XM_011525993.2 XP_011524295.1 14 - 15 1847 82
XM_011525994.2 XP_011524296.1 14 - 15 1847 82
XM_011525998.2 XP_011524300.1 13 - 14 1769 82
XM_011526001.1 XP_011524303.1 12 - 13 1604 82
XM_017025751.1 XP_016881240.1 14 - 15 1802 82
XM_017025752.1 XP_016881241.1 13 - 14 1727 82
XM_017025753.1 XP_016881242.1 13 - 14 1772 82
XM_017025754.1 XP_016881243.1 14 - 15 1802 82
XM_017025755.1 XP_016881244.1 13 - 14 1772 82
XM_017025757.1 XP_016881246.1 14 - 15 1802 82
XM_017025758.1 XP_016881247.1 14 - 15 1802 82
XM_017025759.1 XP_016881248.1 14 - 15 1802 82
XM_017025760.1 XP_016881249.1 13 - 14 1772 82
XM_017025762.1 XP_016881251.1 13 - 14 1727 82
XM_017025763.1 XP_016881252.1 13 - 14 1724 82
XM_017025765.1 XP_016881254.1 12 - 13 1604 82
XM_017025766.1 XP_016881255.1 13 - 14 1679 82
XM_017025767.1 XP_016881256.1 13 - 14 1679 82
XM_017025768.1 XP_016881257.1 12 - 13 1604 82
XM_017025770.1 XP_016881259.1 12 - 13 1604 82
XM_017025771.1 XP_016881260.1 12 - 13 1604 82
XM_017025772.1 XP_016881261.1 12 - 13 1559 82
XM_017025773.1 XP_016881262.1 11 - 12 1535 82
XM_017025774.1 XP_016881263.1 11 - 12 1535 82
GenBank mRNA AF078830.1 - 12 - 13 1588 82
AF078832.1 - 12 - 13 1420 82
AK225889.1 - 13 - 14 1935 82
AK291519.1 - 13 - 14 2026 82
AK295401.1 - 14 - 15 1832 82
AK302004.1 - 14 - 15 1792 82
BC033242.1 - 12 - 13 1583 82
BC047353.1 - 12 - 13 1560 82
Y10746.1 - 12 - 13 1580 82

View Full Product Details