Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MBD1
- Entrez Gene ID
-
4152
- Gene Name
- methyl-CpG binding domain protein 1
- Gene Aliases
-
CXXC3, PCM1, RFT
- Chromosome Location
-
Chr.18: 50266882 - 50281774 on Build GRCh38
View Full Product Details