The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320410.1 | NP_001307339.1 | 1 - 2 | 380 | 80 |
NM_033276.3 | NP_150592.1 | 1 - 2 | 380 | 80 | |
GenBank mRNA | AF078164.2 | - | 1 - 2 | 310 | 80 |
BC012776.2 | - | 1 - 2 | 222 | 80 | |
BC115381.1 | - | 1 - 2 | 221 | 80 | |
BC115382.1 | - | 1 - 2 | 220 | 80 | |
BM557436.1 | - | 1 - 2 | 214 | 80 | |
BU601924.1 | - | 1 - 2 | 383 | 80 |