The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320408.1 | NP_001307337.1 | 3 - 4 | 560 | 83 |
NM_001320409.1 | NP_001307338.1 | 2 - 3 | 474 | 83 | |
NM_033276.3 | NP_150592.1 | 3 - 4 | 520 | 83 | |
XM_011538976.2 | XP_011537278.1 | 2 - 3 | 195 | 83 | |
XM_017020206.1 | XP_016875695.1 | 3 - 4 | 310 | 83 | |
GenBank mRNA | AF078164.2 | - | 3 - 4 | 450 | 83 |
BC012776.2 | - | 3 - 4 | 362 | 83 | |
BC115381.1 | - | 3 - 4 | 361 | 83 | |
BC115382.1 | - | 3 - 4 | 360 | 83 | |
BM544500.1 | - | 3 - 4 | 519 | 83 | |
DA711361.1 | - | 2 - 3 | 437 | 83 |