Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRRM4
Entrez Gene ID
84530
Gene Name
serine/arginine repetitive matrix 4
Gene Aliases
KIAA1853, MU-MB-2.76, nSR100
Chromosome Location
Chr.12: 118981495 - 119163051 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194286.3 NP_919262.2 3 - 4 748 97
GenBank mRNA AB058756.1 - 3 - 4 652 97
AK291361.1 - 3 - 4 748 97
BC152471.1 - 3 - 4 663 97
BC167162.1 - 3 - 4 652 97
BC167163.1 - 3 - 4 652 97
KC995185.1 - 3 - 4 360 97
KC995186.1 - 3 - 4 360 97

View Full Product Details