The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194286.3 | NP_919262.2 | 8 - 9 | 1157 | 82 |
GenBank mRNA | AB058756.1 | - | 8 - 9 | 1061 | 82 |
AK291361.1 | - | 8 - 9 | 1157 | 82 | |
BC152471.1 | - | 8 - 9 | 1072 | 82 | |
BC167162.1 | - | 8 - 9 | 1061 | 82 | |
BC167163.1 | - | 8 - 9 | 1061 | 82 | |
KC995185.1 | - | 8 - 9 | 769 | 82 | |
KC995186.1 | - | 8 - 9 | 769 | 82 |