Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FERMT1
Entrez Gene ID
55612
Gene Name
fermitin family member 1
Gene Aliases
C20orf42, DTGCU2, KIND1, UNC112A, URP1
Chromosome Location
Chr.20: 6074845 - 6123544 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017671.4 NP_060141.3 13 - 14 2504 140
GenBank mRNA AB105105.1 - 13 - 14 2504 140
AF443278.1 - 13 - 14 2073 140
AK000123.1 - 5 - 6 675 140
AK000747.1 - 3 - 4 370 140
AK225216.1 - 5 - 6 675 140
AK314394.1 - 13 - 14 1894 140
AY137240.1 - 12 - 13 1715 140
BC040545.1 - 12 - 13 1759 140
BX647465.1 - 8 - 9 911 141

View Full Product Details